Gene Transcripts

Gene Symbol
WDR19
Entrez Gene ID
57728
Gene Name
WD repeat domain 19
Gene Aliases
ATD5, CED4, DYF-2, IFT144, NPHP13, ORF26, Oseg6, PWDMP, SRTD5
Chromosome Location
Chr. 4: 39182404 - 39285810 on Build GRCh38
UniGene ID
Hs.438482
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001317924.1 NP_001304853.1 6, 7 645
NM_025132.3 NP_079408.3 7, 8 761
XM_011513724.1 XP_011512026.1 7, 8 671
XM_011513725.2 XP_011512027.1 6, 7 626
XM_011513726.2 XP_011512028.1 6, 7 534
XM_011513727.1 XP_011512029.1 5, 6 454
XM_017008501.1 XP_016863990.1 5, 6 454
GenBank mRNA AK026780.1 - 7, 8 636
AK225588.1 - 7, 8 636
AK294730.1 - 6, 7 520
AK304161.1 - 7, 8 634
AK308803.1 - 429
AY029257.1 - 7, 8 761
BC032578.1 - 6, 7 640

View Full Product Details