Gene Transcripts
- Gene Symbol
- C3orf20
- Entrez Gene ID
- 84077
- Gene Name
- chromosome 3 open reading frame 20
- Gene Aliases
- Chromosome Location
- Chr. 3: 14673532 - 14773036 on Build GRCh38
- UniGene ID
- Hs.661452
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_001184957.1 |
NP_001171886.1
|
11, 12 |
1648 |
|
NM_001184958.1 |
NP_001171887.1
|
11, 12 |
1753 |
|
NM_032137.4 |
NP_115513.4
|
11, 12 |
2150 |
|
XM_006713341.2 |
XP_006713404.1
|
11, 12 |
2150 |
|
XM_006713342.2 |
XP_006713405.1
|
11, 12 |
2003 |
|
XM_006713343.2 |
XP_006713406.1
|
11, 12 |
2150 |
|
XM_011534153.2 |
XP_011532455.1
|
11, 12 |
3780 |
|
XM_011534154.2 |
XP_011532456.1
|
11, 12 |
3215 |
|
XM_011534157.2 |
XP_011532459.1
|
7, 8 |
927 |
|
GenBank mRNA |
AK131481.1 |
- |
12, 13 |
2356 |
|
AL136781.1 |
- |
11, 12 |
1568 |
|
AL834386.1 |
- |
11, 12 |
2102 |
|
BC030599.2 |
- |
11, 12 |
1711 |
4831210 |
BC038406.1 |
- |
11, 12 |
2070 |
5167835 |
View Full Product Details