Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_152512.3 | NP_689725.2 | 3 | 706 | |
XM_006724149.3 | XP_006724212.1 | 3 | 1301 | ||
XM_006724150.3 | XP_006724213.1 | 3 | 491 | ||
XM_006724151.1 | XP_006724214.1 | 2 | 195 | ||
XM_006724153.3 | XP_006724216.1 | 3 | 800 | ||
XM_011529927.2 | XP_011528229.1 | 3 | 800 | ||
XM_011529931.2 | XP_011528233.1 | 3 | 800 | ||
GenBank mRNA | BC033895.2 | - | 3 | 706 | |
CR456452.1 | - | 2 | 501 |