Gene Transcripts
- Gene Symbol
- FSIP2
- Entrez Gene ID
- 401024
- Gene Name
- fibrous sheath interacting protein 2
- Gene Aliases
- Chromosome Location
- Chr. 2: 185738895 - 185833289 on Build GRCh38
- UniGene ID
- Hs.98025
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
AK092099.1 |
- |
8 |
1457 |
|
View Full Product Details