Gene Transcripts

Gene Symbol
DNAH12
Entrez Gene ID
201625
Gene Name
dynein axonemal heavy chain 12
Gene Aliases
DHC3, DLP12, DNAH12L, DNAH7L, DNAHC12, DNAHC3, DNHD2, HDHC3, HL-19, HL19
Chromosome Location
Chr. 3: 57293699 - 57545203 on Build GRCh38
UniGene ID
Hs.201378
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001291661.1 NP_001278590.1 56 8982
XM_011533466.1 XP_011531768.1 56 8967
XM_011533467.1 XP_011531769.1 56 8934
XM_011533468.1 XP_011531770.1 56 8904
XM_011533469.1 XP_011531771.1 56 8967
XM_011533471.2 XP_011531773.1 56 8967
XM_017005860.1 XP_016861349.1 56 8967
XM_017005861.1 XP_016861350.1 55 9650
XM_017005862.1 XP_016861351.1 35 5542
GenBank mRNA AK128592.1 - 4 408

View Full Product Details