Gene Transcripts

Gene Symbol
C22orf15
Entrez Gene ID
150248
Gene Name
chromosome 22 open reading frame 15
Gene Aliases
N27C7-3
Chromosome Location
Chr. 22: 23762522 - 23765863 on Build GRCh38
UniGene ID
Hs.116254
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq XM_017028607.1 XP_016884096.1 4 1673
GenBank mRNA AK093880.1 - 3 1525

View Full Product Details