Gene Transcripts

Gene Symbol
HNRNPA3
Entrez Gene ID
220988
Gene Name
heterogeneous nuclear ribonucleoprotein A3
Gene Aliases
2610510D13Rik, D10S102, FBRNP, HNRPA3
Chromosome Location
Chr. 2: 177212694 - 177223959 on Build GRCh38
UniGene ID
Hs.516539
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_194247.2 NP_919223.1 3 411
XM_005246380.1 XP_005246437.1 3 411
XM_005246381.4 XP_005246438.1 3 317
XM_006712365.2 XP_006712428.1 3 411
XM_017003587.1 XP_016859076.1 3 317
XM_017003588.1 XP_016859077.1 3 411
GenBank mRNA AK293125.1 - 3 279
AK304687.1 - 3 274
AK316434.1 - 3 279
AY363225.1 - 3 305
BC064494.1 - 3 323
BC112027.1 - 3 333 8327495
BC113470.1 - 3 333 8322522
BX647217.1 - 3 268
DB093606.1 - 3 411

Gene Symbol
HNRNPA3P1
Entrez Gene ID
10151
Gene Name
heterogeneous nuclear ribonucleoprotein A3 pseudogene 1
Gene Aliases
D10S102, FBRNP, HNRPA3, HNRPA3P1
Chromosome Location
Chr. 10: 43787412 - 43790417 on Build GRCh38
UniGene ID
Hs.632956
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NR_002726.2 1 335

View Full Product Details