Gene Transcripts

Gene Symbol
PRAF2
Entrez Gene ID
11230
Gene Name
PRA1 domain family member 2
Gene Aliases
JM4, Yip6a
Chromosome Location
Chr. X: 49071156 - 49074045 on Build GRCh38
UniGene ID
Hs.29595
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
GenBank mRNA AJ005896.1 - 3 777
AK023433.1 - 3 776
BC021213.2 - 3 763 4297902

View Full Product Details