Gene Transcripts

Gene Symbol
WDR70
Entrez Gene ID
55100
Gene Name
WD repeat domain 70
Gene Aliases
Chromosome Location
Chr. 5: 37379280 - 37752672 on Build GRCh38
UniGene ID
Hs.213690
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_018034.2 NP_060504.1 10, 11 1146
XM_005248318.3 XP_005248375.1 10, 11 1164
XM_011514061.2 XP_011512363.1 4, 5 459
XM_017009604.1 XP_016865093.1 9, 10 980
XM_017009605.1 XP_016865094.1 9, 10 947
GenBank mRNA AK001095.1 - 10, 11 1088
AK291179.1 - 10, 11 1168
AL512685.1 - 10, 11 1147
BC009648.1 - 10, 11 1146 3454326
BC025315.1 - 10, 11 1124 5458850
BX537655.1 - 9, 10 1103

View Full Product Details