Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001282541.1 | NP_001269470.1 | 2 | 339 | |
NM_001282542.1 | NP_001269471.1 | 2 | 339 | ||
NM_003176.3 | NP_003167.2 | 2 | 336 | ||
NR_104211.1 | 2 | 336 | |||
XM_005271155.3 | XP_005271212.1 | 2 | 714 | ||
XM_005271156.1 | XP_005271213.1 | 2 | 299 | ||
XM_006710859.1 | XP_006710922.1 | 2 | 247 | ||
XM_011542037.2 | XP_011540339.1 | 2 | 303 | ||
XM_011542039.1 | XP_011540341.1 | 2 | 303 | ||
XM_017002184.1 | XP_016857673.1 | 2 | 139 | ||
GenBank mRNA | AF273043.1 | - | 2 | 274 | |
AK315268.1 | - | 2 | 298 | ||
BC105299.1 | - | 2 | 229 | ||
BC126266.1 | - | 2 | 213 | 8991982 | |
BC144019.1 | - | 2 | 216 | 9052539 | |
BG772129.1 | - | 2 | 311 | ||
BI561137.1 | - | 2 | 328 | ||
D67035.1 | - | 2 | 272 | ||
X95654.1 | - | 2 | 190 |