Gene Transcripts

Gene Symbol
DCANP1
Entrez Gene ID
140947
Gene Name
dendritic cell associated nuclear protein
Gene Aliases
C5orf20, DCNP1
Chromosome Location
Chr. 5: 135444214 - 135447348 on Build GRCh38
UniGene ID
Hs.152477
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_130848.2 NP_570900.1 1 1461
GenBank mRNA AB074498.1 - 1 1461

View Full Product Details