Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001297608.1 | NP_001284537.1 | 12 | 2736 | |
NM_145263.3 | NP_660306.1 | 13 | 2832 | ||
NR_123728.1 | 13 | 2882 | |||
XM_005265716.1 | XP_005265773.1 | 11 | 2538 | ||
XM_005265717.2 | XP_005265774.1 | 9 | 2204 | ||
XM_005265718.1 | XP_005265775.1 | 9 | 2320 | ||
XM_011534369.1 | XP_011532671.1 | 12 | 2634 | ||
XM_011534370.1 | XP_011532672.1 | 13 | 2758 | ||
XM_011534372.1 | XP_011532674.1 | 10 | 2412 | ||
XM_017007731.1 | XP_016863220.1 | 11 | 2230 | ||
GenBank mRNA | AK304386.1 | - | 12 | 2651 | |
BC025396.1 | - | 13 | 2739 | 4821894 | |
BC037886.1 | - | 7 | 1511 |