Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001145734.2 | NP_001139206.2 | 8 | 1150 | |
NM_001170402.1 | NP_001163873.1 | 8 | 1150 | ||
NM_152623.2 | NP_689836.2 | 8 | 1150 | ||
GenBank mRNA | AB077043.1 | - | 8 | 1119 | |
AB077044.1 | - | 8 | 988 | ||
AB081750.1 | - | 8 | 1119 | ||
AB086378.1 | - | 8 | 1119 | ||
AK095246.1 | - | 8 | 1148 | ||
BC136572.1 | - | 8 | 1020 | 9020562 |