Gene Transcripts

Gene Symbol
ZCCHC12
Entrez Gene ID
170261
Gene Name
zinc finger CCHC-type containing 12
Gene Aliases
PNMA7A, SIZN, SIZN1
Chromosome Location
Chr. X: 118823743 - 118826973 on Build GRCh38
UniGene ID
Hs.21417
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001312891.1 NP_001299820.1 3 1211
NM_173798.3 NP_776159.1 4 1263
GenBank mRNA AK122676.1 - 4 1223
BC031241.1 - 4 1182 5263970
BC036572.2 - 4 1216 5274563
BC057841.1 - 4 1182 5264353

View Full Product Details