Gene Transcripts

Gene Symbol
TSPAN33
Entrez Gene ID
340348
Gene Name
tetraspanin 33
Gene Aliases
PEN, PEN., TSPAN-33
Chromosome Location
Chr. 7: 129144716 - 129169694 on Build GRCh38
UniGene ID
Hs.27267
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_178562.4 NP_848657.1 8 1672
XM_006715960.3 XP_006716023.1 8 1500
GenBank mRNA AK000208.1 - 3 1286
AY236849.1 - 8 1496
BC044244.1 - 8 1636 5759132

View Full Product Details