Gene Transcripts

Gene Symbol
DHX36
Entrez Gene ID
170506
Gene Name
DEAH-box helicase 36
Gene Aliases
DDX36, G4R1, MLEL1, RHAU
Chromosome Location
Chr. 3: 154275668 - 154324497 on Build GRCh38
UniGene ID
Hs.446270
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001114397.1 NP_001107869.1 15 1771
NM_020865.2 NP_065916.2 15 1813
GenBank mRNA AB040921.1 - 13 1265
AF217190.1 - 15 1805
AJ577133.1 - 15 1732
AJ577134.1 - 15 1690
AK303720.1 - 1693
AK314435.1 - 15 1813
BC036035.1 - 15 1777 5273384

View Full Product Details