Gene Transcripts

Gene Symbol
RNF32
Entrez Gene ID
140545
Gene Name
ring finger protein 32
Gene Aliases
FKSG33, HSD15, LMBR2
Chromosome Location
Chr. 7: 156640286 - 156678333 on Build GRCh38
UniGene ID
Hs.446194
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001184996.1 NP_001171925.1 9 1540
NM_001184997.1 NP_001171926.1 9 1393
NM_001308273.1 NP_001295202.1 9 1615
NM_030936.3 NP_112198.1 9 1435
NR_131778.1 9 1389
XM_005249522.4 XP_005249579.1 9 1347
XM_011515804.2 XP_011514106.1 10 1457
XM_011515805.2 XP_011514107.1 10 1614
XM_011515806.2 XP_011514108.1 9 1525
XM_011515807.2 XP_011514109.1 9 1333
XM_011515808.2 XP_011514110.1 9 1445
XM_011515809.2 XP_011514111.1 9 1518
XM_011515810.2 XP_011514112.1 9 1335
XM_011515811.2 XP_011514113.1 9 1330
XM_011515812.2 XP_011514114.1 9 1437
XM_011515813.2 XP_011514115.1 9 1396
GenBank mRNA AF325690.1 - 9 1343
AF441222.1 - 8 1282
AF441224.1 - 9 1389
AL136874.1 - 9 1323

View Full Product Details