Gene Transcripts

Gene Symbol
SLC2A13
Entrez Gene ID
114134
Gene Name
solute carrier family 2 member 13
Gene Aliases
HMIT
Chromosome Location
Chr. 12: 39755021 - 40106085 on Build GRCh38
UniGene ID
Hs.558595
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_052885.3 NP_443117.3 10 2013
XM_011537847.2 XP_011536149.1 9 2113
XM_017018764.1 XP_016874253.1 10 1486
XM_017018765.1 XP_016874254.1 10 2124
XM_017018766.1 XP_016874255.1 9 1271
GenBank mRNA AJ315644.1 - 10 2013
BC117117.1 - 10 1957 40125668
BC117119.1 - 10 1957 40125670

View Full Product Details