Gene Transcripts

Gene Symbol
DCD
Entrez Gene ID
117159
Gene Name
dermcidin
Gene Aliases
AIDD, DCD-1, DSEP, HCAP, PIF
Chromosome Location
Chr. 12: 54644591 - 54648493 on Build GRCh38
UniGene ID
Hs.350570
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001300854.1 NP_001287783.1 6 632
NM_053283.3 NP_444513.1 5 568
GenBank mRNA AF144011.1 - 5 438
BC062682.1 - 5 440 30369929
BC069108.1 - 5 438 7216925
EF503687.1 - 5 568
EF503688.1 - 6 735
EF503689.1 - 6 632

View Full Product Details