Gene Transcripts

Gene Symbol
DUSP15
Entrez Gene ID
128853
Gene Name
dual specificity phosphatase 15
Gene Aliases
C20orf57, VHY
Chromosome Location
Chr. 20: 31845578 - 31870744 on Build GRCh38
UniGene ID
Hs.434108
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001012644.2 NP_001012662.1 7 1076
NM_001320478.1 NP_001307407.1 7 733
NM_080611.4 NP_542178.2 7 1072
NM_177991.2 NP_817130.1 7 838
GenBank mRNA AK091960.1 - 6 1272
BC056911.1 - 7 889 6195838
BM554314.1 - 7 698

View Full Product Details