Gene Transcripts

Gene Symbol
CBLN4
Entrez Gene ID
140689
Gene Name
cerebellin 4 precursor
Gene Aliases
CBLNL1
Chromosome Location
Chr. 20: 55997357 - 56005472 on Build GRCh38
UniGene ID
Hs.126141
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_080617.5 NP_542184.1 3 1742
GenBank mRNA AK289640.1 - 3 1546
AY358527.1 - 3 777
BC050026.1 - 3 1744 4820943
BC069402.1 - 3 441 7262170
BC069488.1 - 3 441 7262182
BC069516.1 - 3 441 7262099

View Full Product Details