Gene Transcripts

Gene Symbol
STRA13
Entrez Gene ID
201254
Gene Name
stimulated by retinoic acid 13
Gene Aliases
CENP-X, CENPX, D9, FAAP10, MHF2
Chromosome Location
Chr. 17: 82018703 - 82022930 on Build GRCh38
UniGene ID
Hs.37616
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001271006.1 NP_001257935.1 5 621
NM_001271007.1 NP_001257936.1 4 552
NM_144998.3 NP_659435.2 4 567
XM_005256339.2 XP_005256396.1 5 773
XM_017024326.1 XP_016879815.1 3 974
XM_017024327.1 XP_016879816.1 4 846
XM_017024328.1 XP_016879817.1 4 715
XM_017024329.1 XP_016879818.1 3 661
GenBank mRNA BC009571.1 - 4 536
BC011610.1 - 4 503
BI837753.1 - 4 535
BQ648002.1 - 1 256
U95006.1 - 4 523
U95007.1 - 4 508

View Full Product Details