Gene Transcripts

Gene Symbol
ZPBP2
Entrez Gene ID
124626
Gene Name
zona pellucida binding protein 2
Gene Aliases
ZPBPL
Chromosome Location
Chr. 17: 39868164 - 39877896 on Build GRCh38
UniGene ID
Hs.367245
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_198844.2 NP_942141.2 3 398
NM_199321.2 NP_955353.1 4 464
XM_011524298.1 XP_011522600.1 4 502
GenBank mRNA AK292383.1 - 4 502
AY251603.1 - 4 447
AY251604.1 - 3 381
BC043152.2 - 3 384 5297840
DB056422.1 - 4 502

View Full Product Details