Gene Transcripts
- Gene Symbol
- ANAPC1
- Entrez Gene ID
- 64682
- Gene Name
- anaphase promoting complex subunit 1
- Gene Aliases
- APC1, MCPR, TSG24
- Chromosome Location
- Chr. 2: 111766147 - 111884196 on Build GRCh38
- UniGene ID
- Hs.436527
- Species
- Homo sapiens
- Gene Symbol
- ANAPC1P1
- Entrez Gene ID
- 100286979
- Gene Name
- anaphase promoting complex subunit 1 pseudogene 1
- Gene Aliases
- Chromosome Location
- Chr. 2: 86861825 - 86895042 on Build GRCh38
- UniGene ID
- Hs.731217
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NR_037931.1 |
|
6 |
813 |
|
GenBank mRNA |
AK310642.1 |
- |
6 |
813 |
|
View Full Product Details