Gene Transcripts

Gene Symbol
SECISBP2
Entrez Gene ID
79048
Gene Name
SECIS binding protein 2
Gene Aliases
SBP2
Chromosome Location
Chr. 9: 89318473 - 89367117 on Build GRCh38
UniGene ID
Hs.59804
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001282688.1 NP_001269617.1 4 632
NM_001282689.1 NP_001269618.1 4 514
NM_001282690.1 NP_001269619.1 4 712
NM_024077.4 NP_076982.3 4 635
XM_005252196.2 XP_005252253.1 4 602
XM_006717282.2 XP_006717345.1 4 568
XM_011519000.2 XP_011517302.1 4 689
XM_011519001.1 XP_011517303.1 4 689
XM_011519002.1 XP_011517304.1 4 689
XM_011519003.1 XP_011517305.1 4 689
XM_017015122.1 XP_016870611.1 4 602
XM_017015123.1 XP_016870612.1 4 602
XM_017015124.1 XP_016870613.1 4 602
XM_017015125.1 XP_016870614.1 2 228
GenBank mRNA AB208940.1 - 4 593
AF380995.1 - 4 588
AK090608.1 - 4 602
AK290182.1 - 4 481
AK299161.1 - 4 723
AK302852.1 - 4 630
AL136881.1 - 4 468
BC001189.1 - 4 468
BC036109.1 - 4 597 5296292
BC068466.1 - 4 570
BC099900.1 - 4 571
BX648787.1 - 2 3575
DR002073.1 - 4 628

View Full Product Details