Gene Transcripts

Gene Symbol
ANO3
Entrez Gene ID
63982
Gene Name
anoctamin 3
Gene Aliases
C11orf25, DYT23, DYT24, GENX-3947, TMEM16C
Chromosome Location
Chr. 11: 26189123 - 26663289 on Build GRCh38
UniGene ID
Hs.577269
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001313726.1 NP_001300655.1 3, 4 482
NM_031418.3 NP_113606.2 2, 3 390
GenBank mRNA AJ300461.1 - 2, 3 1097
AK302659.1 - 2, 3 284
AK304781.1 - 3, 4 323
DA247850.1 - 2, 3 350

View Full Product Details