Gene Transcripts

Gene Symbol
RETNLB
Entrez Gene ID
84666
Gene Name
resistin like beta
Gene Aliases
FIZZ1, FIZZ2, HXCP2, RELM-beta, RELMb, RELMbeta, XCP2
Chromosome Location
Chr. 3: 108755639 - 108757283 on Build GRCh38
UniGene ID
Hs.307047
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_032579.2 NP_115968.1 3 474
GenBank mRNA AF290873.1 - 3 476
AF323084.1 - 3 483
AY358655.1 - 3 475
BC069318.1 - 3 447 7262095
BC113502.1 - 3 408 8322554
BC113528.1 - 3 408 8322580

View Full Product Details