Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | XM_005268208.1 | XP_005268265.1 | 2, 3 | 542 | |
XM_005268209.3 | XP_005268266.1 | 2, 3 | 346 | ||
XM_017021765.1 | XP_016877254.1 | 2, 3 | 345 | ||
GenBank mRNA | AB082526.1 | - | 2, 3 | 514 | |
AK128693.1 | - | 3, 4 | 478 | ||
AY048580.1 | - | 2, 3 | 408 | ||
AY080896.1 | - | 2, 3 | 420 | ||
BC093881.1 | - | 2, 3 | 489 | 7939726 | |
BC112101.1 | - | 2, 3 | 489 | 8327569 |