Gene Transcripts

Gene Symbol
RSPH1
Entrez Gene ID
89765
Gene Name
radial spoke head 1 homolog
Gene Aliases
CT79, RSP44, RSPH10A, TSA2, TSGA2
Chromosome Location
Chr. 21: 42472486 - 42496354 on Build GRCh38
UniGene ID
Hs.661069
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001286506.1 NP_001273435.1 8 986
NM_080860.3 NP_543136.1 9 1100
XM_005261208.2 XP_005261265.1 7 740
XM_011529786.1 XP_011528088.1 8 920
GenBank mRNA AB006536.1 - 9 1100
AB041016.1 - 9 1037
AK057315.1 - 8 2551
BC101519.1 - 9 979 8069025
BC113367.1 - 9 979 8322419

View Full Product Details