Gene Transcripts

Gene Symbol
KBTBD6
Entrez Gene ID
89890
Gene Name
kelch repeat and BTB domain containing 6
Gene Aliases
Chromosome Location
Chr. 13: 41127573 - 41132800 on Build GRCh38
UniGene ID
Hs.534040
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_152903.4 NP_690867.3 1 1731
GenBank mRNA AK056633.1 - 1 1676
AK095195.1 - 1 376
AK096608.1 - 2 1671
AK299627.1 - 2 1479
AL833839.1 - 1 1383
BC000560.2 - 1 1653 3163484
BC051349.1 - 1 1492 6483483

View Full Product Details