Gene Transcripts
- Gene Symbol
- NBEAL1
- Entrez Gene ID
- 65065
- Gene Name
- neurobeachin like 1
- Gene Aliases
- A530083I02Rik, ALS2CR16, ALS2CR17
- Chromosome Location
- Chr. 2: 203014624 - 203225194 on Build GRCh38
- UniGene ID
- Hs.648846
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_001114132.1 |
NP_001107604.1
|
29, 30 |
4973 |
|
XM_005246787.3 |
XP_005246844.1
|
29, 30 |
4999 |
|
XM_005246788.2 |
XP_005246845.1
|
8, 9 |
1475 |
|
XM_006712698.3 |
XP_006712761.1
|
30, 31 |
5086 |
|
XM_006712699.3 |
XP_006712762.1
|
30, 31 |
5086 |
|
XM_011511658.2 |
XP_011509960.1
|
30, 31 |
5041 |
|
XM_011511659.2 |
XP_011509961.1
|
30, 31 |
5098 |
|
XM_011511660.2 |
XP_011509962.1
|
26, 27 |
4420 |
|
XM_011511662.2 |
XP_011509964.1
|
22, 23 |
3755 |
|
XM_011511663.2 |
XP_011509965.1
|
30, 31 |
5076 |
|
XM_017004728.1 |
XP_016860217.1
|
17, 18 |
3153 |
|
XM_017004729.1 |
XP_016860218.1
|
5, 6 |
1167 |
|
GenBank mRNA |
AK127772.1 |
- |
3, 4 |
841 |
|
View Full Product Details