Gene Transcripts

Gene Symbol
KCTD9
Entrez Gene ID
54793
Gene Name
potassium channel tetramerization domain containing 9
Gene Aliases
BTBD27
Chromosome Location
Chr. 8: 25424357 - 25458468 on Build GRCh38
UniGene ID
Hs.188763
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_017634.3 NP_060104.2 4 464
GenBank mRNA AK298518.1 - 4 268
BC021216.2 - 4 267 4302317
BC068518.1 - 4 277 5265435
BG249272.1 - 398
BU149129.1 - 4 382
DA115324.1 - 4 465

View Full Product Details