Gene Transcripts
- Gene Symbol
- BEST2
- Entrez Gene ID
- 54831
- Gene Name
- bestrophin 2
- Gene Aliases
- VMD2L1
- Chromosome Location
- Chr. 19: 12751691 - 12758458 on Build GRCh38
- UniGene ID
- Hs.435611
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
AK000139.1 |
- |
9 |
1552 |
|
AK225232.1 |
- |
9 |
1552 |
|
View Full Product Details