Gene Transcripts

Gene Symbol
SBNO1
Entrez Gene ID
55206
Gene Name
strawberry notch homolog 1
Gene Aliases
MOP3, Sno
Chromosome Location
Chr. 12: 123289109 - 123365204 on Build GRCh38
UniGene ID
Hs.7012
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001167856.1 NP_001161328.1 6 890
NM_018183.3 NP_060653.3 6 887
XM_005253572.3 XP_005253629.1 7 1028
XM_005253573.4 XP_005253630.1 8 1438
XM_005253575.3 XP_005253632.1 7 1025
XM_005253576.3 XP_005253633.1 8 1069
XM_006719473.3 XP_006719536.1 9 1194
XM_006719474.3 XP_006719537.1 8 1147
XM_011538533.2 XP_011536835.1 9 1188
XM_011538534.2 XP_011536836.1 6 926
XM_011538535.2 XP_011536837.1 6 928
XM_017019556.1 XP_016875045.1 7 1012
XM_017019557.1 XP_016875046.1 7 1019
XM_017019558.1 XP_016875047.1 8 1082
XM_017019559.1 XP_016875048.1 7 1149
GenBank mRNA AB014772.1 - 6 887
AK001695.1 - 7 1025
AK122703.1 - 8 1443
AK307488.1 - 6 926
AY364255.1 - 6 889
BC030544.1 - 7 1021
BC133704.1 - 6 890 40124902

View Full Product Details