Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_018328.4 | NP_060798.2 | 7 | 1149 | |
XM_011511470.2 | XP_011509772.1 | 7 | 1381 | ||
XM_011511472.2 | XP_011509774.1 | 5 | 961 | ||
XM_017004482.1 | XP_016859971.1 | 3 | 831 | ||
XM_017004483.1 | XP_016859972.1 | 3 | 831 | ||
XM_017004484.1 | XP_016859973.1 | 3 | 831 | ||
XM_017004485.1 | XP_016859974.1 | 3 | 831 | ||
GenBank mRNA | AB040894.1 | - | 2 | 442 | |
AK304424.1 | - | 2 | 464 | ||
BC150264.1 | - | 2 | 442 | 9007212 | |
EF542797.1 | - | 7 | 1140 |