Gene Transcripts
- Gene Symbol
- HIST1H2AB
- Entrez Gene ID
- 8335
- Gene Name
- histone cluster 1, H2ab
- Gene Aliases
- H2A/m, H2AFM
- Chromosome Location
- Chr. 6: 26033092 - 26033568 on Build GRCh38
- UniGene ID
- Hs.248174
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_003513.2 |
NP_003504.2
|
1 |
284 |
|
View Full Product Details