Gene Transcripts

Gene Symbol
FEM1A
Entrez Gene ID
55527
Gene Name
fem-1 homolog A
Gene Aliases
EPRAP
Chromosome Location
Chr. 19: 4791716 - 4795559 on Build GRCh38
UniGene ID
Hs.515082
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_018708.2 NP_061178.1 1 2580
GenBank mRNA AJ311359.1 - 1 2440
AK074232.1 - 1 2538
AK126532.1 - 2 2308
AL359589.1 - 1 718
AL365408.1 - 1 1035
AY356352.1 - 1 2534
BC004988.1 - 1 2538 2957950
CR749732.1 - 1 718

View Full Product Details