Gene Transcripts

Gene Symbol
ASAH2
Entrez Gene ID
56624
Gene Name
N-acylsphingosine amidohydrolase 2
Gene Aliases
BCDase, HNAC1, LCDase, N-CDase, NCDase
Chromosome Location
Chr. 10: 50184861 - 50251564 on Build GRCh38
UniGene ID
Hs.512645
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001143974.1 NP_001137446.1 3 415
NM_019893.2 NP_063946.2 3 415
XM_011539970.2 XP_011538272.1 4 611
XM_011539971.2 XP_011538273.1 4 615
XM_011539972.2 XP_011538274.1 4 612
XM_011539973.2 XP_011538275.1 4 621
XM_017016435.1 XP_016871924.1 4 615
GenBank mRNA AF250847.1 - 3 358
AF449759.2 - 3 415
BC107105.2 - 3 358 40018929

View Full Product Details