Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_020648.5 | NP_065699.1 | 5 | 1125 | |
GenBank mRNA | AF196834.1 | - | 5 | 1082 | |
AF294628.1 | - | 4 | 948 | ||
AK027629.1 | - | 7 | 1283 | ||
AK075550.1 | - | 5 | 1125 | ||
BC020490.1 | - | 5 | 1039 | 3889103 |