Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001290185.1 | NP_001277114.1 | 2 | 539 | |
NM_021729.5 | NP_068375.3 | 2 | 324 | ||
XM_011542899.2 | XP_011541201.1 | 2 | 282 | ||
GenBank mRNA | AB027508.2 | - | 2 | 294 | |
AF308800.1 | - | 2 | 258 | ||
AK023279.1 | - | 2 | 296 | ||
AK056701.1 | - | 2 | 287 | ||
AK302973.1 | - | 2 | 471 | ||
AL834218.1 | - | 2 | 273 | ||
BC065563.1 | - | 2 | 258 | 6144541 | |
BI561271.1 | - | 2 | 329 |