Gene Transcripts

Gene Symbol
SAV1
Entrez Gene ID
60485
Gene Name
salvador family WW domain containing protein 1
Gene Aliases
SAV, WW45, WWP4
Chromosome Location
Chr. 14: 50633106 - 50668353 on Build GRCh38
UniGene ID
Hs.642842
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_021818.3 NP_068590.1 5 1472
XM_011537057.2 XP_011535359.1 6 1535
XM_017021568.1 XP_016877057.1 6 1765
GenBank mRNA AJ292969.1 - 5 1251
AK021500.1 - 5 1419
AK023071.1 - 5 1301
AK095903.1 - 6 1336
AK225896.1 - 5 1444
AK290883.1 - 6 1344
AL833378.1 - 4 882
BC020537.1 - 5 1418 4425620
CR457297.1 - 5 1086

View Full Product Details