Gene Transcripts

Gene Symbol
HIST1H2AM
Entrez Gene ID
8336
Gene Name
histone cluster 1, H2am
Gene Aliases
H2A.1, H2A/n, H2AFN, dJ193B12.1
Chromosome Location
Chr. 6: 27892699 - 27893185 on Build GRCh38
UniGene ID
Hs.134999
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_003514.2 NP_003505.1 1 438
GenBank mRNA BC032686.1 - 1 453
BC032756.1 - 1 453

View Full Product Details