Gene Transcripts

Gene Symbol
LMOD3
Entrez Gene ID
56203
Gene Name
leiomodin 3
Gene Aliases
NEM10
Chromosome Location
Chr. 3: 69108672 - 69122595 on Build GRCh38
UniGene ID
Hs.350621
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001304418.1 NP_001291347.1 3 1061
NM_198271.4 NP_938012.2 2 1155
GenBank mRNA AK057852.1 - 2 682
AK300096.1 - 3 1034
AL832033.1 - 2 1126
AL832709.1 - 2 1160
BC121019.2 - 2 1034 40117347

View Full Product Details