Gene Transcripts

Gene Symbol
DNAJB5
Entrez Gene ID
25822
Gene Name
DnaJ heat shock protein family (Hsp40) member B5
Gene Aliases
Hsc40
Chromosome Location
Chr. 9: 34989369 - 34998433 on Build GRCh38
UniGene ID
Hs.237506
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001135004.2 NP_001128476.2 5 1329
NM_001135005.2 NP_001128477.1 4 1536
NM_012266.5 NP_036398.3 4 1184
XM_005251428.3 XP_005251485.1 4 1103
XM_006716751.2 XP_006716814.1 5 1193
XM_011517842.2 XP_011516144.1 5 1770
GenBank mRNA AF087870.1 - 5 1186
AF088982.1 - 4 1087
AK023253.1 - 4 1080
AK299647.1 - 4 1536
AK314671.1 - 4 1061

View Full Product Details