Gene Transcripts

Gene Symbol
OSTM1
Entrez Gene ID
28962
Gene Name
osteopetrosis associated transmembrane protein 1
Gene Aliases
GIPN, GL, HSPC019, OPTB5
Chromosome Location
Chr. 6: 108041409 - 108074737 on Build GRCh38
UniGene ID
Hs.226780
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_014028.3 NP_054747.2 2, 3 595
GenBank mRNA AF533891.1 - 2, 3 579
AK075012.1 - 2, 3 521
AK129854.1 - 2, 3 589
AK314393.1 - 2, 3 271
AY358795.1 - 2, 3 586
BC068581.1 - 2, 3 577 5298450
BI668275.1 - 2, 3 611

View Full Product Details