Gene Transcripts

Gene Symbol
R3HCC1L
Entrez Gene ID
27291
Gene Name
R3H domain and coiled-coil containing 1 like
Gene Aliases
C10orf28, GIDRP86, GIDRP88, PSORT
Chromosome Location
Chr. 10: 98134608 - 98244897 on Build GRCh38
UniGene ID
Hs.419800
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001256619.1 NP_001243548.1 5 1046
NM_001256620.1 NP_001243549.1 4 995
NM_014472.4 NP_055287.4 4 991
NM_138469.2 NP_612478.2 5 1088
XM_011539639.1 XP_011537941.1 5 1303
XM_011539640.1 XP_011537942.1 6 1108
XM_011539641.1 XP_011537943.1 5 1060
XM_011539642.1 XP_011537944.1 6 1400
XM_011539643.1 XP_011537945.1 4 987
XM_011539644.1 XP_011537946.1 4 991
XM_011539645.2 XP_011537947.1 4 1234
XM_011539646.1 XP_011537948.1 3 866
XM_011539647.1 XP_011537949.1 3 868
XM_011539648.1 XP_011537950.1 2 741
XM_011539649.1 XP_011537951.1 5 1303
XM_017016074.1 XP_016871563.1 4 2199
XM_017016075.1 XP_016871564.1 2 741
XM_017016076.1 XP_016871565.1 5 1035
XM_017016077.1 XP_016871566.1 7 1198
XM_017016078.1 XP_016871567.1 6 1120
XM_017016079.1 XP_016871568.1 6 1134
XM_017016080.1 XP_016871569.1 4 1040
XM_017016081.1 XP_016871570.1 5 1122
XM_017016082.1 XP_016871571.1 3 914
XM_017016083.1 XP_016871572.1 4 930
GenBank mRNA AF050198.1 - 4 983
AF525304.1 - 5 999
AK292546.1 - 4 990
BC098268.1 - 5 1029 40026913
BC098359.1 - 4 932 40026912
BC099848.1 - 4 932 40026916

View Full Product Details