Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001009182.1 | NP_001009182.1 | 5 | 457 | |
NM_001009183.1 | NP_001009183.1 | 5 | 457 | ||
NM_003616.2 | NP_003607.1 | 5 | 457 | ||
XM_017021709.1 | XP_016877198.1 | 5 | 480 | ||
GenBank mRNA | AB037701.1 | - | 5 | 435 | |
AB037702.1 | - | 5 | 435 | ||
AB037703.1 | - | 5 | 355 | ||
AF027150.1 | - | 5 | 518 | ||
AK313947.1 | - | 5 | 445 | ||
BC028095.1 | - | 6 | 574 | ||
BC104968.1 | - | 5 | 456 | 8143971 | |
CN484054.1 | - | 5 | 457 |