Gene Transcripts
- Gene Symbol
- GOLM1
- Entrez Gene ID
- 51280
- Gene Name
- golgi membrane protein 1
- Gene Aliases
- C9orf155, GOLPH2, GP73, HEL46, PSEC0257, bA379P1.3
- Chromosome Location
- Chr. 9: 86026141 - 86100201 on Build GRCh38
- UniGene ID
- Hs.494337
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_016548.3 |
NP_057632.2
|
3, 4 |
517 |
|
NM_177937.2 |
NP_808800.1
|
3, 4 |
509 |
|
GenBank mRNA |
AF236056.1 |
- |
3, 4 |
458 |
|
AK027717.1 |
- |
3, 4 |
480 |
|
AK075542.1 |
- |
3, 4 |
451 |
|
AK075557.1 |
- |
3, 4 |
421 |
|
AK092943.1 |
- |
4, 5 |
397 |
|
AY358593.1 |
- |
3, 4 |
422 |
|
BC001740.1 |
- |
3, 4 |
432 |
3351316 |
BQ216766.1 |
- |
|
476 |
|
CR457201.1 |
- |
2, 3 |
308 |
|
DA116177.1 |
- |
3, 4 |
510 |
|
EU794616.1 |
- |
3, 4 |
476 |
|
View Full Product Details