Gene Transcripts

Gene Symbol
TIMM17A
Entrez Gene ID
10440
Gene Name
translocase of inner mitochondrial membrane 17 homolog A (yeast)
Gene Aliases
TIM17, TIM17A
Chromosome Location
Chr. 1: 201955491 - 201970661 on Build GRCh38
UniGene ID
Hs.20716
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_006335.2 NP_006326.1 6 1163
GenBank mRNA AF106622.1 - 6 1133
AK023063.1 - 3 2337
BC007106.1 - 6 1144 4284092
BC009784.1 - 6 1155 3683125
BC013823.1 - 6 1122
BC015098.1 - 6 1161 3686369
BC016817.1 - 4 1019
BC020833.1 - 6 1141 4773149

View Full Product Details