Gene Transcripts

Gene Symbol
ZER1
Entrez Gene ID
10444
Gene Name
zyg-11 related cell cycle regulator
Gene Aliases
C9orf60, ZYG, ZYG11BL
Chromosome Location
Chr. 9: 128729786 - 128772507 on Build GRCh38
UniGene ID
Hs.147950
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_006336.3 NP_006327.2 13 2462
XM_005251645.2 XP_005251702.1 13 2254
XM_005251646.2 XP_005251703.1 13 2451
XM_011518122.2 XP_011516424.1 13 2264
XM_017014185.1 XP_016869674.1 13 2870
XM_017014186.1 XP_016869675.1 13 2245
XM_017014187.1 XP_016869676.1 13 2864
XM_017014188.1 XP_016869677.1 13 2232
GenBank mRNA AK314911.1 - 13 2210
BC001447.2 - 2 257
BC052563.1 - 13 2447 6303965
X99802.1 - 12 2052

View Full Product Details